Cleanroom tape 0160WH200-P3S, vinyl tape with rubber adhesive, medium adhesion for strong bond and easy release. Width 2 (51mm), length 36 yds (33m), color white. Classic clean version, for use in ISO 7/8 cleanrooms, 1 roll per single bag. 1 * 13 Rolle
UTAP0160WH200-P3S
Frequently Bought Together