ContactUsLinkComponent
Troemner OIML Precision Weights Gewicht, 50mg, OIML E2, ZERT 1 * 1 ST
Catalog # OHAU80780289
Supplier:  OHAUS
Troemner OIML Precision Weights Gewicht, 50mg, OIML E2, ZERT 1 * 1 ST
Catalog # OHAU80780289
Supplier:  OHAUS
Frequently Bought Together