ContactUsLinkComponent
Frequency converter for stepless speed adjustment from 23 - 101 min-1 with potentiometer, speed measuring with digital display and timer with ON/OFF switch for setting mixing time. The electrical controls are in a separate housing, splash water proof 1 * 1 ST
Artikelnr. IBSAFREQCONVERTER
Lieferant:  IBS Stock Items
Frequency converter for stepless speed adjustment from 23 - 101 min-1 with potentiometer, speed measuring with digital display and timer with ON/OFF switch for setting mixing time. The electrical controls are in a separate housing, splash water proof 1 * 1 ST
Artikelnr. IBSAFREQCONVERTER
Lieferant:  IBS Stock Items
Frequently Bought Together