ContactUsLinkComponent
Piperacillin / tazobactam 30/6 mcg Disc . Each package should include 50 disc and each Disc should include piperacillin/tazobactam 30/6 mcg 1 * 250 ST
Catalog # HIMESD292E-1PK
Supplier:  HIMEDIA
Piperacillin / tazobactam 30/6 mcg Disc . Each package should include 50 disc and each Disc should include piperacillin/tazobactam 30/6 mcg 1 * 250 ST
Catalog # HIMESD292E-1PK
Supplier:  HIMEDIA
Frequently Bought Together