Cefepime 30 mcg Disc. Each package should include 50 disc and each disk should include 30 mcg cefepime 1 * 250 ST
Artikelnr. HIMESD219-1PK
Lieferant: HIMEDIA
Cefepime 30 mcg Disc. Each package should include 50 disc and each disk should include 30 mcg cefepime 1 * 250 ST
Artikelnr. HIMESD219-1PK
Lieferant: HIMEDIA
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Lieferant: VWR Chemicals
Sealing film for manual application, Parafilm® M
Lieferant: HEATHROW
VWR® Gesichtstücher
Lieferant: VWR Collection
VWR®, Nitrilhandschuhe
Lieferant: VWR Collection
Acetonitril ≥99,9%, HiPerSolv CHROMANORM® für die LC-MS, geeignet für UPLC/UHPLC-Instrumente
Lieferant: VWR Chemicals
pH-Indikatorstäbchen, MQuant®, Supelco®
Lieferant: Merck
Methanol ≥99,8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Gradienten-Qualität für die HPLC
Lieferant: VWR Chemicals
Wasser, HiPerSolv CHROMANORM® für die HPLC
Lieferant: VWR Chemicals
VWR® NITRILE LIGHT, Nitrilhandschuhe
Lieferant: VWR Collection
VWR®, Transferpipetten, High Performance
Lieferant: VWR Collection
Nitrilhandschuhe, Kimtech™ Purple Nitrile™
Lieferant: KIMBERLY CLARK
VWR®, Serologische Pipetten, Standard Serie
Lieferant: VWR Collection