AuftragsverfolgungKontakt
MultiRack racked tips, Axygen® | Avantor
MultiRack racked tips, Axygen®
Standard length, 1 - 300 µl, +
Artikelnr. AXYGMR-300-R-S
Anbieter:   Corning
MultiRack racked tips, Axygen®
Artikelnr. AXYGMR-300-R-S
Anbieter:  Corning
MultiRack racked tips, Axygen®
MultiRack racked tips, Axygen®
Standard length, 1 - 300 µl, +
Preis
346,00 €
Einloggen für Verfügbarkeit
1 * 4.800 ST
Product Details & Documents
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Non pyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
Axygen® MultiRack tips are supplied in convenient hinged racks that help reduce airborne contamination of the tips during use. Each rack is individually sealed. The high-quality polypropylene racks are reusable and fully autoclavable. They can be refilled with Axygen® MultiRack reload systems to save both money and plastic. Sturdy racks resist accidental tipping, and the easy-close lid lock prevents tips falling while closed. The transparent rack and color-coded decks allow easy size identification. Maximum recovery are also available.
Ordering information: Axygen® MultiRack pipette tips are also available in bulk, filter tips, reload, and tipstation configurations.
Spezifikationen
  • Steril:
    +
  • Volumen:
    1 - 300 µl
  • Seq:
    06
  • Spitzentyp:
    Standard length