AuftragsverfolgungKontakt
Multirack bulk tips, Axygen® | Avantor
Multirack bulk tips, Axygen®
Extended length, 0,1 - 10 µl
Artikelnr. AXYGMR-10XT
Anbieter:   Corning
Multirack bulk tips, Axygen®
Artikelnr. AXYGMR-10XT
Anbieter:  Corning
Multirack bulk tips, Axygen®
Multirack bulk tips, Axygen®
Extended length, 0,1 - 10 µl
Preis
399,00 €
Einloggen für Verfügbarkeit
1 * 10.000 ST
Product Details & Documents
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an inter-changeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Nonpyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
Axygen® MultiRack bulk pipette tips are compatible with MultiRack racks and packaged in resealable polyethylene bags, non sterile.
 : Axygen® MultiRack pipette tips are also available in hinged rack, filter tips, reload, and tipstation configurations.
Spezifikationen
  • Volumen:
    0,1 - 10 µl
  • Seq:
    01
  • Spitzentyp:
    Extended length