VISCOSITY STANDARD SILICON 251.5 MPA.S 1 * 600 mL
PARGVIS-RT250-600
Frequently Bought Together
Leitfähigkeitsstandardlösungen
: VWR Chemicals
VWR® AVS TITRINORM Pufferlösungen, 20 °C
: VWR Chemicals
Buffer solutions
: Mettler - Toledo
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
: MERCK PRODUCTION CHEMICALS
Primärfarblösungen, Reag. Ph. Eur.
: VWR Chemicals