VISCOSITY STANDARD GENERAL 72328 MPA.S 1 * 500 mL
PARGS30000
Frequently Bought Together
VWR® AVS TITRINORM Pufferlösungen, 20 °C
: VWR Chemicals
Leitfähigkeitsstandardlösungen
: VWR Chemicals
VWR® Pufferlösungen, Sekundärstandards
: VWR Chemicals
pH buffer solutions
: Mettler - Toledo
Buffer solutions
: Mettler - Toledo