ContactUsLinkComponent
VISCOSITY STANDARD GENERAL 392 MPA.S 1 * 500 mL
Artikelnr. PARGS200
Lieferant:  PARAGON SCIENTIFIC
VISCOSITY STANDARD GENERAL 392 MPA.S 1 * 500 mL
Artikelnr. PARGS200
Lieferant:  PARAGON SCIENTIFIC
Frequently Bought Together