Genuine Tecan® 300 mL Reagent Trough or Reservoir , flat bottom, suitable for Tecan® Liquid Handling Arm™ (LiHa), Flexible Channel A rm (FCA) and MultiChannel Arm™(MCA). 40 troughs/sales unit. 1 * 40 ST
Artikelnr. TCAN30077312
Lieferant: Tecan
Frequently Bought Together