Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Nylon Wire and Cable Ties

Nylon cable ties, 3,5 mm, Gelb, 20 cm
  BRDY196352
 :   Brady
Nylon Wire and Cable Ties
  BRDY196352
 :  Brady
Nylon Wire and Cable Ties
Nylon Wire and Cable Ties

Nylon cable ties, 3,5 mm, Gelb, 20 cm

 
9,60 €
   
1 * 100 ST
 
 
  • Farbe:
    Gelb
  • Bezeichnung:
    Nylon cable ties, 3,5 mm
  • Länge:
    20 cm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.