Nylon Wire and Cable Ties
Nylon cable ties, width 3,6 mm, Gelb, 14 cm
Artikelnr. BRDY196346
Anbieter: Brady
Springe zu:
Product Details & Documents
Spezifikationen
- Farbe:Gelb
- Bezeichnung:Nylon cable ties, width 3,6 mm
- Länge:14 cm
- Seq:04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



