ContactUsLinkComponent
Stainless Steel Shear Beam Load Cell - W & M approval Class III, 2x3000d (=0,02%) acc. to OIML - nominal load: 454 kg - max. usable load 200% of nominal load - version with threaded hole - protection class: IP 67 - cabel: 3 m, in 6-wire technology, s 1 * 1 ST
Artikelnr. IBSAMP58/454KG
Lieferant:  IBS Stock Items
Stainless Steel Shear Beam Load Cell - W & M approval Class III, 2x3000d (=0,02%) acc. to OIML - nominal load: 454 kg - max. usable load 200% of nominal load - version with threaded hole - protection class: IP 67 - cabel: 3 m, in 6-wire technology, s 1 * 1 ST
Artikelnr. IBSAMP58/454KG
Lieferant:  IBS Stock Items
Frequently Bought Together