ContactUsLinkComponent
Recombinant Mouse Lipoprotein lipase(Lpl), 0.05mg (E.coli) Expression Region: 28-474aa Full Length of Mature Protein Tag information: Tag-Free Sequence: ADAGRDFSDIESKFALRTPEDTAEDTCHLIPGLADSVSNCHFNHSSKTFVVIHGWTVTGMYESWVPKLVAAL YKREPDSNVIVVDWLYRAQQHYP 1 * 0,05 mg
Artikelnr. MSPPMBS7114946
Recombinant Mouse Lipoprotein lipase(Lpl), 0.05mg (E.coli) Expression Region: 28-474aa Full Length of Mature Protein Tag information: Tag-Free Sequence: ADAGRDFSDIESKFALRTPEDTAEDTCHLIPGLADSVSNCHFNHSSKTFVVIHGWTVTGMYESWVPKLVAAL YKREPDSNVIVVDWLYRAQQHYP 1 * 0,05 mg
Artikelnr. MSPPMBS7114946
Frequently Bought Together