ContactUsLinkComponent
Green dye to stain live and dead bacteria ?Ex/?Em (with DNA) = 500/520 nm Detect in the FITC channel for flow cytometry and microscope 1 * 100 µl
Artikelnr. 40102.
Lieferant:  Biotium
Green dye to stain live and dead bacteria ?Ex/?Em (with DNA) = 500/520 nm Detect in the FITC channel for flow cytometry and microscope 1 * 100 µl
Artikelnr. 40102.
Lieferant:  Biotium
Frequently Bought Together