AuftragsverfolgungKontakt
FLED fluorescence intermediate attachments | Avantor
FLED fluorescence intermediate attachments
FLED Fluorescence intermediate attachment (TRITC)
- Fluorescence intermediate part
- LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
- Light blocking slider
Artikelnr. MOTI1101011400201
Anbieter:   MOTIC
FLED fluorescence intermediate attachments
Artikelnr. MOTI1101011400201
Anbieter:  MOTIC
FLED fluorescence intermediate attachments
FLED fluorescence intermediate attachments
FLED Fluorescence intermediate attachment (TRITC)
- Fluorescence intermediate part
- LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
- Light blocking slider
Preis
1 950,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Compatible with Panthera U, KU and CC microscopes.
Spezifikationen
  • Bezeichnung:
    FLED Fluorescence intermediate attachment (TRITC)
    - Fluorescence intermediate part
    - LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
    - Light blocking slider