Human CD16a (V176), His Tag (SPR BLI verified) (CD8-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession P08637-1). 1 * 1 mg
Artikelnr. ACRBCD8-H52H4-1MG
Lieferant: ACRO Biosystems
Frequently Bought Together