Bell-Flo Complete,500mL-70mm The Bell-Flo Spinner Flask utilizes a rugged stainless steel PTFE internal bearing assembly. The flask comes with either 32mm or 45 mm side arm capsfor port assemblies. Bottom indents with three vertical baffles for optim 1 * 1 ST
BELC1965-80575
Frequently Bought Together