ContactUsLinkComponent
9 Pos Process Stirr Comp,230V Specifications: Number of positions: Nine (9) Max. Flask Size: 500 ml (max. of 9 vessels with 70mm necks) **Larger vessels may be stirred if fewer positions are used** Speed Range: 15-500 rpm Dimensions: 16.5 x 18 x 3 3 1 * 1 ST
Artikelnr. BELC7785-P9015
Lieferant:  BELLCO GLASS
9 Pos Process Stirr Comp,230V Specifications: Number of positions: Nine (9) Max. Flask Size: 500 ml (max. of 9 vessels with 70mm necks) **Larger vessels may be stirred if fewer positions are used** Speed Range: 15-500 rpm Dimensions: 16.5 x 18 x 3 3 1 * 1 ST
Artikelnr. BELC7785-P9015
Lieferant:  BELLCO GLASS
Frequently Bought Together