ContactUsLinkComponent
Human CD155, Fc Tag (CD5-H5251) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accession NP006496.2). 1 * 1 mg
Artikelnr. ACRBCD5-H5251-1MG
Lieferant:  ACRO Biosystems
Human CD155, Fc Tag (CD5-H5251) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accession NP006496.2). 1 * 1 mg
Artikelnr. ACRBCD5-H5251-1MG
Lieferant:  ACRO Biosystems
Frequently Bought Together