Submersible pump, set till 90 m, Teflon Submersible pump set, for purging and sampling monitoring wells to a depth of 90 m and inside diameter from 50 mm. Standard set with submersible pump Grundfos MP 1, converter, Teflon tube, cables, cooling jacke 1 * 1 ST
Artikelnr. EIJK12.27SH
Lieferant: EIJKELKAMP AGRISECTOR
Frequently Bought Together