Biological indicators, strips
STRIP CONTROL BAT E6: Paper strips inoculated with spores of Bacillus atropheus (ATCC® 9372) (1 - 5×106 CFU/strip) and tubes of nutrient broth with pH indicator, 100 strips
Supplier: LIOFILCHEM
Jump to:
Product Details & Documents
Biological indicators for validating the sterilisation using steam sterilisation (autoclaves).
Specifications
- Bezeichnung:STRIP CONTROL BAT E6: Paper strips inoculated with spores of Bacillus atropheus (ATCC® 9372) (1 - 5×106 CFU/strip) and tubes of nutrient broth with pH indicator, 100 strips