Express Eingabe
Austria
AuftragsverfolgungKontakt
Biological indicators, strips
Biological indicators, strips
STRIP CONTROL BAT E6: Paper strips inoculated with spores of Bacillus atropheus (ATCC® 9372) (1 - 5×106 CFU/strip) and tubes of nutrient broth with pH indicator, 100 strips
Catalog # 91067.
Supplier:   LIOFILCHEM
Biological indicators, strips
Catalog # 91067.
Supplier:  LIOFILCHEM
Biological indicators, strips
STRIP CONTROL BAT E6: Paper strips inoculated with spores of Bacillus atropheus (ATCC® 9372) (1 - 5×106 CFU/strip) and tubes of nutrient broth with pH indicator, 100 strips
Price
1 020,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Biological indicators for validating the sterilisation using steam sterilisation (autoclaves).
Specifications
  • Bezeichnung:
    STRIP CONTROL BAT E6: Paper strips inoculated with spores of Bacillus atropheus (ATCC® 9372) (1 - 5×106 CFU/strip) and tubes of nutrient broth with pH indicator, 100 strips
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.