ContactUsLinkComponent
Human CD32b, His Tag (SPR  BLI verified) (CDB-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 217 (Ac cession P31994). 1 * 1 mg
Artikelnr. ACRBCDB-H5228-1MG
Lieferant:  ACRO Biosystems
Human CD32b, His Tag (SPR  BLI verified) (CDB-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 217 (Ac cession P31994). 1 * 1 mg
Artikelnr. ACRBCDB-H5228-1MG
Lieferant:  ACRO Biosystems
Frequently Bought Together