ContactUsLinkComponent
LCO/42H/DIG ~ OVEN 1250 litres capacity Temperature range 60 to 250°C, fluctuation ± 0.5°C Aluminium coated mild steel interior Easy clean powder coated body Chamber dimensions (cms): 142 high x 122 wide x 74 deep External dimensions (cms): 178 high 1 * 1 ST
Artikelnr. GENLLCO/42H/DIG240
Lieferant:  GENLAB
LCO/42H/DIG ~ OVEN 1250 litres capacity Temperature range 60 to 250°C, fluctuation ± 0.5°C Aluminium coated mild steel interior Easy clean powder coated body Chamber dimensions (cms): 142 high x 122 wide x 74 deep External dimensions (cms): 178 high 1 * 1 ST
Artikelnr. GENLLCO/42H/DIG240
Lieferant:  GENLAB
Frequently Bought Together