Waring MX1200XTXEES Standmixer . Variable-speed control dial operates from 1,500 to 24,000RPM PULSE feature overrides speed 10 at 30,000+RPM . 220-240V 50-60Hz 1500 W 1,000 RPM (Low) 20,000 RPM (high) 27,00 RPM (Pulse) 6,5 AMPS CE . Same as MX1200XTX 1 * 1 ST
Artikelnr. WARIMX1200XTXEES
Lieferant: WARING BLENDOR
Frequently Bought Together