Spheroid microplates, Ultra-Low Attachment surface, Corning®
Round bottom, spheroid with lid, ultra-low attachment surface, 1536, Schwarz/klar, 12 μl, +
Anbieter: Corning
Spheroid microplates, Ultra-Low Attachment surface, Corning®
Artikelnr. 738-0218
Anbieter: Corning
Springe zu:
Product Details & Documents
PS, black opaque microplate with optically clear bottom, with lid. The Ultra-Low Attachment surface is a covalently bound hydrogel layer that is hydrophilic and neutrally charged. The surface minimises cell attachment, protein absorption, and enzyme activation. It is ideal for culturing stem cells, as it promotes embryoid body formation from ES cells.
- Well geometry aids formation of spheroids in centre of well
- Non cytotoxic, biologically inert and non degradable surface
- Sterilised by gamma radiation
Zertifikate
Suche nach ChargennummerA Novel Method for Generating Single, Intestinal Organoids for High Throughput Screening A Novel Three Dimensional Immune Oncology Model for High Throughput Testing of Tumoricidal Capability Co-culturing and Assaying Spheroids in the Corning® Spheroid Microplate Considerations for three-dimensional cell culture when using the Corning® spheroid microplate Corning spheroid microplates Spheroid formation protocol
Mehr
Spezifikationen
- Anz. Wells:1536
- Steril:+
- Farbe:Schwarz/klar
- Bezeichnung:Round bottom, spheroid with lid, ultra-low attachment surface
- Seq:05
- Wellvolumen:12 μl
- Cat. No.:738-0218
- Umweltzertifizierung:RECs, ISO 14001 - Kennebunk
- Umweltfreundlichkeit:Umweltschonende Herstellung
Complementary Products
Produktinformationen
- Artikelnr.ModellAuflösungBezeichnungVEFarbeWellvolumenSterilAnz. WellsVerpacktKulturflächeVerfügbarkeitPreis
- MLDVMICRO4ImageXpress Micro 425 - 100 nmHigh-Content-Imaging-SystemEinloggen für Verfügbarkeit1 * 1 ST - 0,00 €
- 735-0364MIKROKONFOKAL25 nmHigh-content mikrokonfokales BildgebungssystemEinloggen für Verfügbarkeit1 * 1 ST - 0,00 €
- MLDVNANOMXImage Xpress NanoImaging-System mit MetaXpress-Software001Einloggen für Verfügbarkeit1 * 1 ST - 0,00 €
- 734-1100Corning® Matrigel® Basalmembranmatrix0210 mLEinloggen für Verfügbarkeit1 * 10 mL - 536,08 €
- 734-2763Schwarz/klar200 µl+Half area microplate, with lid, tissue culture treated01Einloggen für Verfügbarkeit1 * 16 ST - 389,00 €
- 734-2971Einzeln verpackt113 cm²Standard-12-Well-Platte, 6× oberflächenbehandelte Scaffold-Einsätze (21,0×1,6 mm)Einloggen für Verfügbarkeit1 * 8 ST - 287,00 €
- 392-0608Klar400 µl+MicroWell™ 96-Well-Platte, flacher Boden, Nunclon™ Sphera™ behandelte Oberfläche06Einloggen für Verfügbarkeit1 * 8 ST - 222,00 €



