0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Securibox-Behälter PP weiss, 180 ml 1 * 100 ST
SEMA7690
:
SEMADENI
Securibox-Behälter PP weiss, 180 ml 1 * 100 ST
SEMA7690
:
SEMADENI
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Kryoboxen, halbe Größe, 136x66,5 mm
: VWR Collection
VWR®, Flaschen zur Wasserprobenahme, steril
: VWR Collection
Kryoboxen, 133×133 mm
: RATIOLAB
Serologische Pipetten, Stripette™
: Corning
Kryoboxen, B69/B50/B80/B99-Serie
: GLW
VWR®, Kryoboxen, Karton, 133×133 mm
: VWR Collection
Petrischalen
: Greiner Bio-One
VWR®, Transferpipetten, High Performance
: VWR Collection
Vial, flip-top
: Capitol Europe