Grinding Ball 6,35 mm Stainless Steel Lysing Matrix Tubes are ideal for grinding, lysis and homogenization of your most resistant samples The stainless steel threaded cap provides a leak-proof closure without the energy-robbing alternatives like pla 1 * 6 ST
Artikelnr. 116991006.
Lieferant: MP Biomedicals
Frequently Bought Together