Objectives for REVOLVE 3M microscope
Plan achromat; 0,1 NA, 4×, 18,5 mm
Catalog # ECHLJMAG0044
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
ECHO Meet Revolve
Product Literature
Specifications
- Vergrößerung:4×
- Bezeichnung:Plan achromat; 0,1 NA
- Seq:04
- Arbeitsabstand:18,5 mm