ContactUsLinkComponent
Kit, RAEGuard 2 PID, ATEX/IECEx, 1 - 1000 ppm isobutylene equivalent Note: Correction factors and relays are not available for this detection range 1 * 1 ST
Catalog # RAESH-D03-0008-000
Supplier:  RAE SYSTEMS
Kit, RAEGuard 2 PID, ATEX/IECEx, 1 - 1000 ppm isobutylene equivalent Note: Correction factors and relays are not available for this detection range 1 * 1 ST
Catalog # RAESH-D03-0008-000
Supplier:  RAE SYSTEMS
Frequently Bought Together