ContactUsLinkComponent
Kit for Illumina® für 96 Reaktionen - Perform fragmentation, end repair and dA-tailing with a single enzyme mix - Experience reliable fragmentation with a single protocol, regardless of DNA input amount or GC content - Prepare high quality libraries 1 * 1 KIT
Artikelnr. NEWE7805L
Lieferant:  NEW ENGLAND BIOLABS
Kit for Illumina® für 96 Reaktionen - Perform fragmentation, end repair and dA-tailing with a single enzyme mix - Experience reliable fragmentation with a single protocol, regardless of DNA input amount or GC content - Prepare high quality libraries 1 * 1 KIT
Artikelnr. NEWE7805L
Lieferant:  NEW ENGLAND BIOLABS
Frequently Bought Together