NucleoMag DNA FFPE (4x96) 4x 96 preps for the purification of genomic DNA from FFPE samples - NucleoMag B-Beads, buffers, Proteinase K __UN 1993 Entzündbare Flüssigkeit, n.a.g. (Ethanol-Mischung) 3 III 0,800 L ADR/GGVS 1 * 4 ST
Catalog # MANA744320.4
Supplier: MACHEREY-NAGEL
Frequently Bought Together