Porablot NCP (0.3 x 3 m, 1 roll) nitrocellulose membrane, pore size: 0.45 µm, size: 0.3 x 3 m pack of 1 roll __UN 3270 Membranfil ter aus Nitrocellulose 4.1 II 0,17 Kg ADR/GGVSEB F1 LQ PAX 458 1 Kg CAO 458 15 Kg Q: 0,17 / 0, 1 * 1 Rolle
Artikelnr. MANA741280
Lieferant: MACHEREY-NAGEL
Frequently Bought Together