NucleoSpin 96 DNA Plasma (4x96) 4 x 96 preps for the isolation of cell-free DNA from blood plasma - NucleoSpin 96 Plasma Binding Pla te, Buffers, Proteinase K , 24-Square-well Blocks, Round-well Blocks, NucleoSpin 96 Plasma Binding Plates __UN 1993 1 * 384 ST
Artikelnr. MANA740873.4
Lieferant: MACHEREY-NAGEL
Frequently Bought Together