NucleoBond Xtra BAC Buffer Set buffer set for the isolation of large constructs, like BACs, PACs, and P1, only applicable with Nucle oBond Xtra BAC kits - Buffer RES-BAC, Buffer LYS-BAC, Buffer NEU-BAC (1000 mL each), RNase A kein Gefahrgut nach Tra 1 * 1 ST
Catalog # MANA740437
Supplier: MACHEREY-NAGEL
Frequently Bought Together