ContactUsLinkComponent
NucleoBond Xtra EF Buffer Set I buffer set for the isolation of endotoxin-free low-copy plasmids, BACs, PACs, and P1 constructs, onl y applicable with NucleoBond Xtra EF kits, sufficient for 12 Xtra Maxi preps or 18 Xtra Midi preps - Buffer RES-EF, B 1 * 1 SET
Artikelnr. MANA740427
Lieferant:  MACHEREY-NAGEL
undefined
NucleoBond Xtra EF Buffer Set I buffer set for the isolation of endotoxin-free low-copy plasmids, BACs, PACs, and P1 constructs, onl y applicable with NucleoBond Xtra EF kits, sufficient for 12 Xtra Maxi preps or 18 Xtra Midi preps - Buffer RES-EF, B 1 * 1 SET
Artikelnr. MANA740427
Lieferant:  MACHEREY-NAGEL
Frequently Bought Together