ContactUsLinkComponent
NucleoBond Xtra Buffer Set I buffer set for the isolation of low-copy plasmids, BACs, PACs, and P1 constructs, only applicable with NucleoBond Xtra kits, sufficient for 12 Xtra Maxi preps or 18 Xtra Midi preps - Buffer RES, Buffer LYS, Buffer NEU, RN 1 * 1 SET
Artikelnr. MANA740417
Lieferant:  MACHEREY-NAGEL
undefined
NucleoBond Xtra Buffer Set I buffer set for the isolation of low-copy plasmids, BACs, PACs, and P1 constructs, only applicable with NucleoBond Xtra kits, sufficient for 12 Xtra Maxi preps or 18 Xtra Midi preps - Buffer RES, Buffer LYS, Buffer NEU, RN 1 * 1 SET
Artikelnr. MANA740417
Lieferant:  MACHEREY-NAGEL
Frequently Bought Together