NucleoVac Valves (24) Valves for handling different flow rates of NucleoSpin and NucleoSnap columns on a NucleoVac 24 Vacuum Manifol d Pack of 24 1 * 24 ST
NucleoVac Valves (24) Valves for handling different flow rates of NucleoSpin and NucleoSnap columns on a NucleoVac 24 Vacuum Manifol d Pack of 24 1 * 24 ST