This product was prepared from normal serum by delipidation, selective precipitation and tandem molecular sieve chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipi 1 * 1 mg
Artikelnr. AVIVOPRA03402
Lieferant: Aviva Systems Biology
Frequently Bought Together