Express Eingabe
Austria
AuftragsverfolgungKontakt
Kits for Precellys®
Kits for Precellys®
Empty metal tubes set for Precellys® Evolution homogenisers, 6×7 ml metal tubes, tube holder and blocking plate, Personalised filling
Artikelnr. 432-0306
Anbieter:   Bertin Instruments
Kits for Precellys®
Artikelnr. 432-0306
Anbieter:  Bertin Instruments
Kits for Precellys®
Kits for Precellys®
Empty metal tubes set for Precellys® Evolution homogenisers, 6×7 ml metal tubes, tube holder and blocking plate, Personalised filling
Preis
2 510,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
The Precellys lysing kits offer the best versatility of volumes and grinding material to process any type of samples: Human, animal, plants, microorganisms or soil.
  • More than 30 different lysing matrixes
  • Reinforced tubes are suitable for use at the maximum energy levels
  • Easy-handling features allow the Precellys to be used without any advanced training
These kits are specifically for sample preparation for Precellys homogenisers. They can process any type of sample. Choosing the appropriate lysing kit maximises the quantity and quality of extraction by decreasing the process time and limiting the heat generated.
Reinforced tubes have thicker walls and their top contains an o-ring to make it more resistant.
Spezifikationen
  • Bezeichnung:
    Empty metal tubes set for Precellys® Evolution homogenisers, 6×7 ml metal tubes, tube holder and blocking plate
  • Seq:
    055
  • Für:
    Personalised filling
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.