XCE pan-cen is composed of orange labeled repetitive sequences specific for human centromeres except for chromosome 1 which is ident ified by repetitive sequences specific for the heterochromatic region 1q12. 1 * 0,5 mL
Artikelnr. MSPPD-0836-050-OR
Lieferant: MARKET SOURCE PART PROCESS
Frequently Bought Together