4'-Hydroxy-3'-methoxyacetophenon
Bottle, 25 g
Artikelnr. APOSOR26691-25G
Anbieter: Apollo Scientific
4'-Hydroxy-3'-methoxyacetophenon
Artikelnr. APOSOR26691-25G
Anbieter: Apollo Scientific
CAS Nummer: 498-02-2
Springe zu:
Product Details & Documents
Synonyme: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin
- CAS Nummer:498-02-2
- Formel:C₉H₁₀O₃
- MDL-Nummer:MFCD00008747
- Molekulargewicht:166.18 g/mol
- Siedepunkt:263…265 °C
- Schmelzpunkt:112…115 °C
- Blitzpunkt:113 °C
- Lagertemperatur:Raumtemperatur
Spezifikationen
Spezifikationen
- Packung:Bottle
- VE:25 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



