Folsäure ≥98%
Bottle, 100 g
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
- CAS Number:59-30-3
- Formula:C₁₉H₁₉N₇O₆
- MDL Number:MFCD00079305
- Molecular Weight:441.41 g/mol
- Storage Temperature:Raumtemperatur
- EINECS:200-419-0
Certificates
Search by Lot NumberSpecifications
Specifications
- Packung:Bottle
- VE:100 g



