Express Eingabe
Austria
AuftragsverfolgungKontakt
AMPSO ≥98%
AMPSO ≥98%
Bottle, 100 g
  APOSBID0501-100G
AMPSO ≥98%
  APOSBID0501-100G
 :  68399-79-1
AMPSO ≥98%
Bottle, 100 g
 :  68399-79-1
262,00 €
1 * 100 g
Biological buffer - useful range 8.3-9.7.
 :  N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulphonic acid
  •  :68399-79-1
  •  :C₇H₁₇NO₅S
  •  :MFCD00041777
  •  :227.28 g/mol
  •  :Raumtemperatur
  • Packung:
    Bottle
  • VE:
    100 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.