AMPSO ≥98%
Bottle, 100 g
APOSBID0501-100G
Biological buffer - useful range 8.3-9.7.
: N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulphonic acid
- :68399-79-1
- :C₇H₁₇NO₅S
- :MFCD00041777
- :227.28 g/mol
- :Raumtemperatur
- Packung:Bottle
- VE:100 g