(±)-α-Bromphenylessigsäure
Bottle, 100 g
Artikelnr. APOSOR350067-100G
Anbieter: Apollo Scientific
(±)-α-Bromphenylessigsäure
Artikelnr. APOSOR350067-100G
Anbieter: Apollo Scientific
CAS Nummer: 4870-65-9
Springe zu:
Product Details & Documents
Synonyme: α-Bromophenylacetic Acid, DL-α-Bromophenylacetic acid
- CAS Nummer:4870-65-9
- Formel:C₈H₇BrO₂
- MDL-Nummer:MFCD00004206
- Molekulargewicht:215.0440 g/mol
- Schmelzpunkt:82…83 °C
- Lagertemperatur:Raumtemperatur
- UN:3261
- ADR:8,II
Spezifikationen
Spezifikationen
- Packung:Bottle
- VE:100 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



