Pasteur pipettes
Graduated, 1 ml, 35 µl, 150 mm, +, Individually wrapped, 500/carton
Artikelnr. GREI740365_500
Anbieter: Greiner Bio-One
Springe zu:
Product Details & Documents
These pipettes are made from PE.
Spezifikationen
- Verpackt:Individually wrapped, 500/carton
- Steril:+
- Kapazität:1 ml
- Bezeichnung:Graduated
- Länge:150 mm
- Seq:03
- Tropfen-Volumen:35 µl