AuftragsverfolgungKontakt
96-Well plate sample control system, U-2D™
Inserts, clear borocilicate glass, 500 µl, for use with U-2D™ rack
Catalog # MICS95296-I-500
Supplier:   MICROSOLV
96-Well plate sample control system, U-2D™
Catalog # MICS95296-I-500
Supplier:  MICROSOLV
96-Well plate sample control system, U-2D™
96-Well plate sample control system, U-2D™
Inserts, clear borocilicate glass, 500 µl, for use with U-2D™ rack
Price
114,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
The U-2D™ universal two dimensional 96-well plate system offer users unprecedented sample management control compared to current glass lined or plastic well plates for sample preparation or chromatography.
Easily push up the insert from the bottom and remove it from the rack. These inserts can then be placed into any MicroSolv 9 mm screw top autosampler vial for safe storage, transport or for a second chromatography dimension, or to another instrument platform.
 : Base and sealing mats sold separately.
Specifications
  • Bezeichnung:
    Inserts, clear borocilicate glass, 500 µl, for use with U-2D™ rack
  • Seq:
    05
Accessories
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • MICS95296-PDL-PK
    Paddles for U-2D™ sealing film
    Log in for availability
    1 * 5 ST - 17,10 €
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • MICS95296-ROLLER
    Roller for U-2D™ sealing film
    Log in for availability
    1 * 1 ST - 65,60 €
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • MICS95296-U2D
    U-2D™ plate, 350µL, with rack and base only
    Log in for availability
    1 * 1 ST - 116,00 €
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • MICS95296-ZFF-PK
    Zone free sealing film for U-2D™ plates
    Log in for availability
    1 * 10 ST - 54,50 €
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • MICS95296-ZFF-CS
    Zone free sealing film for U-2D™ plates
    Log in for availability
    1 * 50 ST - 228,00 €