ContactUsLinkComponent
Stand U with Articulated Suspension Arm (D) - Table attachment device U 210 mm - Table clamp U (5...75 mm table thickness) - Articulated suspension arm U 580 mm with rotary knob for height locking (lifting range 500 mm, carrying capacity 2…4.8 Kg) - 1 * 1 ST
Artikelnr. ZEIS4354149000000
Lieferant:  ZEISS
Stand U with Articulated Suspension Arm (D) - Table attachment device U 210 mm - Table clamp U (5...75 mm table thickness) - Articulated suspension arm U 580 mm with rotary knob for height locking (lifting range 500 mm, carrying capacity 2…4.8 Kg) - 1 * 1 ST
Artikelnr. ZEIS4354149000000
Lieferant:  ZEISS
Frequently Bought Together