Capillary installation gauges
Capillary installation gauge for Thermo Scientific TRACE and Focus SSL (M4 ferrules)
Catalog # REST22330
Supplier: Restek
Jump to:
Product Details & Documents
These capillary installation gauges for Agilent GCs are made from high-quality stainless steel.
- Prevents crushed column ends
- Seats graphite ferrules onto column for consistent installations
Specifications
- Bezeichnung:Capillary installation gauge for Thermo Scientific TRACE and Focus SSL (M4 ferrules)
- Seq:04