Express Eingabe
Austria
AuftragsverfolgungKontakt
Capillary installation gauges
Capillary installation gauges
Capillary installation gauge for Thermo Scientific TRACE and Focus SSL (M4 ferrules)
Catalog # REST22330
Supplier:   Restek
Capillary installation gauges
Catalog # REST22330
Supplier:  Restek
Capillary installation gauges
Capillary installation gauges
Capillary installation gauge for Thermo Scientific TRACE and Focus SSL (M4 ferrules)
Price
119,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These capillary installation gauges for Agilent GCs are made from high-quality stainless steel.
  • Prevents crushed column ends
  • Seats graphite ferrules onto column for consistent installations
Specifications
  • Bezeichnung:
    Capillary installation gauge for Thermo Scientific TRACE and Focus SSL (M4 ferrules)
  • Seq:
    04